Demonstration Pulleys
Demonstration Pulleys, Triple
Catalog # 470235-700
Fournisseur #: 4219-00-10
Jump to:
Discontinued
Product Details & Documents
Jumbo-Sized Pulleys are Designed for Classroom Demonstrations
These pulleys are open to make it easier for users to run pulley cords through them. The single, double, and triple pulleys are all 2 3/4" in diameter. Light-weight, low-friction, and deformation-resistant plastic ensures accurate results.
Spécifications
- Désignation:Demonstration Pulleys, Triple