Fermentation Tubes
Fermentation Tube, Ungraduated, 5 mL, with Foot
470235-690
: 601-9-10
Verify gas production in fermentation exercises, gas produced by yeast or other organisms rises into a 5 or 10 ml evolution tube.
- Ungraduated with foot
- Désignation:Fermentation Tube, Ungraduated, 5 mL, with Foot