$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
BIN STACK/HANG BLACK 5-3/8 X 4-1/8 X 3IN
76202-728
:
QUANTUM STORAGE SYSTEMS
BIN STACK/HANG BLACK 5-3/8 X 4-1/8 X 3IN
76202-728
:
QUANTUM STORAGE SYSTEMS
:
QUS210BK
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International