biOrb® Maintenance Materials
Polish And Cloth Accessory
Catalog # 470234-882
biOrb® Maintenance Materials
Catalog # 470234-882
Supplier:  
biOrb® Maintenance Materials
biOrb® Maintenance Materials
Polish And Cloth Accessory
Discontinued
Product Details & Documents
Keep Your biOrb® Working Like New
  • Original maintenance materials
  • Maintenance for the inside and outside
These biOrb® maintenance materials are perfect for maintaining all biOrb® aquarium products. As regular maintenance all biOrb® aquaria require regular filter changes about every 4 to 8 weeks. The service kits contain all you need for normal filter changes, a new cartridge, a special cleaning pad for the interior of the aquarium, and instructions. The Marine service kit also includes some reef grade salt and water supplements to balance your water after a change. If you like to stock ahead, the Service pack X3 includes all you need to make 3 filter changes, and also includes a bottle of water optimizer.
Is the exterior of your biOrb® looking like it needs some help. The acrylic aquaria doesn't need much care, but over time you want to clean it up. In order to keep your biOrb® looking its best, the manufacturer recommends using these cleaners, polishes and cleaning cloths on the acrylic finish.
Spécifications
  • Désignation:
    Polish And Cloth Accessory