SoftMax Pro® 7.X Data Acquisition and Analysis Software, Molecular Devices
Non-expiring SoftMax Pro® 7 license, Four activiation keys
  CA75838-332
 :  SMP7PROF
SoftMax Pro® 7.X Data Acquisition and Analysis Software, Molecular Devices
  CA75838-332
SoftMax Pro® 7.X Data Acquisition and Analysis Software, Molecular Devices
SoftMax Pro® 7.X Data Acquisition and Analysis Software, Molecular Devices
Non-expiring SoftMax Pro® 7 license, Four activiation keys
$10,703.77
Each
SoftMax® Pro 7 Software is the full solution, from data acquisition to analysis to publishing.
  • Recommended Computer Configuration for SoftMax® Pro 7.X:
  • Windows 7 (32-bit or 64-bit), Windows 8 (32-bit or 64-bit), or Windows 10 (32-bit or 64-bit)
  • Quad core CPU
  • USB 2.0 ports
  • 4 GB RAM
  • At least 1 GB of free hard disk space
  • 32-bit graphics display with 256 MB video RAM
  • 1280x1024 or higher-resolution display
Read data using over 160 ready-to-run experimental protocols from one of 23 Molecular Devices microplate readers, including FlexStation® 3, or import from another data source using the new SoftMax Pro® Import Feature, which is available as an add-on to SoftMax Pro® 6.4.1 or higher.
The inclusive software package includes 21 curve fit options, advanced analysis algorithms, and custom reporting tools to analyze and present data in the best possible way. Whether quantifying nucleic acids and proteins or setting up a compound screen to determine IC50 values and relative potency by parallel line analysis, SoftMax Pro® provides the simplicity needed for new users as well as the refined computational control needed for complex data analysis, LIMS, and robotics integration.
 : FlexStation® 2, SpectraMax® L, LMax I & II, THERMOmax, UVmax, and SpectraMax® 250 are not supported by SoftMax Pro® 7.X.
  • Désignation:
    Non-expiring SoftMax Pro® 7 license
  • Includes:
    Four activiation keys