Paper Circuits Kit | Avantor
Paper Circuits Kit
Standard kit, 10
Catalog # 470234-652
Supplier:   BROWN DOG GADGETS
Fournisseur #:  K1006-STA
Paper Circuits Kit
Catalog # 470234-652
Supplier:  BROWN DOG GADGETS
Paper Circuits Kit
Paper Circuits Kit
Standard kit, 10
Price
44,10 $
Log in pour la disponibilité
Each
Product Details & Documents
Perfect for putting the 'A' in STEAM, why mess with wires when you can use conductive tape?
  • Includes jumbo LEDs and conductive maker tape to make your project shine!
  • Perfect for makers of all ages
  • No soldering or special tools needed
Paper circuits are made with LEDs, coin cell batteries, conductive maker tape, and everyday craft supplies. Use templates available online or create your own designs to light up a craft, poster or tri-fold presentation board.
Paper circuits are economical, engaging and a great tool for hands-on learning of simple circuitry. The standard kit contains materials for at least 10 projects. The classroom kit contains materials for at least 25 projects.
Ordering information: Standard kit includes (30) jumbo LEDs; (12) binder clips; (10) CR2032 coin cell batteries; (1) roll of ¹/₄"×5 m maker tape; access to online resources.
Classroom kit includes (120) jumbo LEDs; (40) binder clips; (40) CR2032 coin cell batteries; (2) rolls of ¹/₄"×20 m maker tape; access to online resources.
Spécifications
  • Désignation:
    Standard kit
  • No. of Students:
    10
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Conductive Maker Tape™, ¼" × 20 m
    Conductive Maker Tape™, ¼" × 20 m
    470234-666
    Log in pour la disponibilité
    Each - 16,45 $
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Conductive maker tape, ⅛" × 5 m
    Conductive maker tape, ⅛" × 5 m
    470234-664
    Log in pour la disponibilité
    Each - 8,70 $