Prepared Agarose
Prepared Agarose Gel, 0.8%, 200 ml Bottle
: AA0084-200ML
Prepared with TBE buffer, these gels are easy to use — simply melt them in a microwave oven or water bath then pour them into a casting tray. The non-reusable gels have a one-year shelf life and should be stored at room temperature.
- Désignation:Prepared Agarose Gel, 0.8%, 200 ml Bottle