Prepared Agarose
Prepared Agarose Gel, 0.8%, 200 ml Bottle
  470233-562
 :  AA0084-200ML
Prepared Agarose
  470233-562
Prepared Agarose
Prepared Agarose
Prepared Agarose Gel, 0.8%, 200 ml Bottle
$33.00
Each (200ml)
Prepared with TBE buffer, these gels are easy to use — simply melt them in a microwave oven or water bath then pour them into a casting tray. The non-reusable gels have a one-year shelf life and should be stored at room temperature.
  • Désignation:
    Prepared Agarose Gel, 0.8%, 200 ml Bottle