Somso® Catheterization Demonstration Models | Avantor
Somso® Catheterization Demonstration Models
Catheterization-Model, Female
Catalog # 470233-262
Supplier:   MARCUS SOMMER
Fournisseur #:  CLA7/14
Somso® Catheterization Demonstration Models
Catalog # 470233-262
Supplier:  MARCUS SOMMER
Somso® Catheterization Demonstration Models
Somso® Catheterization Demonstration Models
Catheterization-Model, Female
Price
$462.00
Log in pour la disponibilité
Each
Product Details & Documents
Show your students what's happening.
  • Great for basic practice
  • Opens to show catheter in place
  • Male and Female versions available
Ideal for demonstrating disposable and balloon catheters as well as suprapubic aspiration. Can be dismantled into two parts to show the catheter in position. In natural size, made of special plastic. On a stand with green base.
Spécifications
  • Désignation:
    Catheterization-Model, Female
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.