Somso® Catheterization Demonstration Models
Catheterization-Model, Female
Catalog # 470233-262
Supplier: MARCUS SOMMER
Fournisseur #: CLA7/14
Jump to:
Product Details & Documents
Show your students what's happening.
- Great for basic practice
- Opens to show catheter in place
- Male and Female versions available
Ideal for demonstrating disposable and balloon catheters as well as suprapubic aspiration. Can be dismantled into two parts to show the catheter in position. In natural size, made of special plastic. On a stand with green base.
Spécifications
- Désignation:Catheterization-Model, Female