Wallcur® PRACTI- Labels | Avantor
Wallcur® PRACTI- Labels
PRACTI-Isoproterenol, 0.2 mg/ml
Catalog # 470233-116
Supplier:   WALLCUR, LLC.
Fournisseur #:  9905SUP
Wallcur® PRACTI- Labels
Catalog # 470233-116
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI- Labels
Wallcur® PRACTI- Labels
PRACTI-Isoproterenol, 0.2 mg/ml
Price
$32.09
Log in pour la disponibilité
Pack of 100
Product Details & Documents
Label Your Existing Vials!
  • Pre-Printed Labels Showing Name And Concentrations
  • Appropriately Sized For The Most Common Containers Used
  • Contains Warnings Calling Out That The Contents Are For Training Only
Customize your labs ability to train using these peel and stick labels. These are the same labels used to label the PRACTI- line of simulated products. Each pack contains 100 labels so you can adjust the mix of options available in your lab. Each label also contains a barcode, so you can incorporate individual vials into an inventory management system for training.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Spécifications
  • Désignation:
    PRACTI-Isoproterenol
  • Taille:
    0.2 mg/ml