Wallcur® PRACTI-Vials | Avantor
Wallcur® PRACTI-Vials
Practi-10 ml tint vial, Distilled water
Catalog # 470232-836
Supplier:   WALLCUR, LLC.
Fournisseur #:  422TV
Wallcur® PRACTI-Vials
Catalog # 470232-836
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI-Vials
Wallcur® PRACTI-Vials
Practi-10 ml tint vial, Distilled water
Price
$117.49
Log in pour la disponibilité
Box of 30
Product Details & Documents
Have A Variety On Hand For Skills Training
  • Contains No Medication
  • Prefilled With Distilled Water
  • Clear And Tinted Vials Available
  • Safe For Use With Most Simulators
Teach your students the skills necessary for using all sizes of vials. These generically labeled vials are pre-filled with distilled water, so are safe to use with most simulators and injection trainers. Early training with generic vials will build confidence in your students as they prepare to work with specifically labeled vials latter in your nursing, EMS, or Pharm tech training program.
These Vials are great for teaching the following:
Small or large vial handling dexterity techniques
Aseptic precautions specific to specifically sized vials
Air pressure changes / air replacement technique
Multiple small dose calculations
Fractional dosage measurement
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Spécifications
  • Désignation:
    Practi-10 ml tint vial
  • Includes:
    Distilled water