Wallcur® PRACTI-Ampules | Avantor
Wallcur® PRACTI-Ampules
PRACTI-1 ml Mini Tint Amp, Distilled Water
Catalog # 470232-818
Supplier:   WALLCUR, LLC.
Fournisseur #:  413MT
Wallcur® PRACTI-Ampules
Catalog # 470232-818
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI-Ampules
Wallcur® PRACTI-Ampules
PRACTI-1 ml Mini Tint Amp, Distilled Water
Price
$165.79
Log in pour la disponibilité
Box of 100
Product Details & Documents
Prefilled And Etched For Proper Breakage.
  • Contains No Medication
  • Contains Distilled Water, Suitable For Use In Simulators And Injection Pads
  • Available Unlabeled, Or Labeled
  • Amber And Clear Versions Available To Simulate Light Sensitive Medications
Your students will learn how to accurately open, calculate, aspirate, and measure small and often hard to see solutions. Wallcur ampules are chemically scored for ease of use and to accurately simulate those found in hospitals and clinics. Unlabeled ampules can be combined with the PRACTI-Dose labels for additional medication simulations.
Optimizes Proficiency in:
Fluid displacement from top to bottom
Ampule opening/breaking
Sterile technique during aspiration
Dosage measurement
Informations sur les accessoires: For safer practice pair your ampule with the PRACTI-Ampule Safety Breaker.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Spécifications
  • Désignation:
    PRACTI-1 ml Mini Tint Amp
  • Includes:
    Distilled Water