Wallcur® PRACTI-Insulin
Wallcur® PRACTI-Insulin
PRACTI-Insulin Variety Pack 10 NPH, 10 Reg, 1070/30, 10 Lantuz, Barium Sulfate And Water
Catalog # 470232-810
Supplier:   WALLCUR, LLC.
Supplier #:  408VP
Wallcur® PRACTI-Insulin
Catalog # 470232-810
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI-Insulin
Wallcur® PRACTI-Insulin
PRACTI-Insulin Variety Pack 10 NPH, 10 Reg, 1070/30, 10 Lantuz, Barium Sulfate And Water
Price
134,49 $
Log in for availability
Kit of 1
Product Details & Documents
Practice With A Wide Variety Of Insulin Products
  • Contains No Medication
  • Multiple Options Available
Teaching insulin injections starts with the fundamentals of carefully reading the insulin vial labels for dosage information as many insulin labels look alike. With PRACTI- Insulin you can teach your students how to calculate dosages, choose the applicable syringe for the dosage, draw up the correct amount of air, and then rotate, mix and withdraw before the medication precipitates out. Wallcur’s PRACTI- Insulin Vials contain distilled water and an additive that closely simulates the real thing and must be mixed properly, and administered promptly.
Teach your students to:
Identify and understand two of the most commonly ordered insulin medications
Read and interpret differing insulin labels that often look-a-like
Calculate insulin dosages
Mix, combine, and promptly withdraw short and long acting insulin
Identify injection site landmarks, and aseptic precautions when administering insulin
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Specifications
  • Désignation:
    PRACTI-Insulin Variety Pack 10 NPH, 10 Reg, 1070/30, 10 Lantuz
  • Includes:
    Barium Sulfate And Water