Fine Mesh Basket, Sklar® | Avantor
Fine Mesh Basket, Sklar®
Fine Mesh Basket with Locking Clip, Stainless steel, 3×11/2×3/4"
Catalog # 76191-842
Supplier:   Sklar
Fournisseur #:  10-6612
Fine Mesh Basket, Sklar®
Catalog # 76191-842
Supplier:  Sklar
Fine Mesh Basket, Sklar®
Fine Mesh Basket, Sklar®
Fine Mesh Basket with Locking Clip, Stainless steel, 3×11/2×3/4"
Price
$388.54
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
Spécifications
  • Désignation:
    Fine Mesh Basket with Locking Clip
  • Dimensions:
    3×11/2×3/4"
  • Matière:
    Stainless steel