Utility Scissors, OR-Grade, Sklar® | Avantor
Utility Scissors, OR-Grade, Sklar®
7.5", Stainless Steel/Plastic
Catalog # 76191-650
Supplier:   Sklar
Fournisseur #:  11-1285
Utility Scissors, OR-Grade, Sklar®
Catalog # 76191-650
Supplier:  Sklar
Utility Scissors, OR-Grade, Sklar®
Utility Scissors, OR-Grade, Sklar®
7.5", Stainless Steel/Plastic
Price
$62.80
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting.
Spécifications
  • Longueur:
    7.5"
  • Matière:
    Stainless Steel/Plastic