Ramelet Phlebectomy Hook, Sklar®
Product Details & Documents
Sklar's® Ramelet Phlebectomy Hook is used to grip the vein during surgical procedures.
- With industrial standard
- With five major surgical instruments line
- Delivers superior precision for any surgical procedure
The hook comes in fine and medium tips, angled left and right. The handle is round and knurled, which makes it easier to roll between the fingers for better control.
Spécifications
- Désignation:Ramelet Phlebectomy Hook, Medium
- Longueur:10.2 cm (4")
- Matière:Stainless steel
- Curvature:Angled (Right)