Ward's® Meniscus Reader
Lecteur de ménisque
Catalog # 470232-208
Supplier: VWR International
Jump to:
Product Details & Documents
Easily see the endpoint of your titration.
Provides background that is white above and black beneath
- Transparent front strap holds reader in place
- Waterproof - flexible plastic card, not laminated paper
The Ward's® Meniscus reader makes it vastly easier to visualize the bottom of the meniscus in burets and small graduated cylinders. The contrasting black and white background allows students to easily see the meniscus in the buret - AND accurately read the volume. Slide the meniscus reader up or down to position the black demarcation just below the meniscus.
Specifications
- Désignation:Lecteur de ménisque