Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Computer/Technology Tables, All Welded, FlipTop, AmTab

Flip top computer and technology table, 4 - 5, 29", 84×30×29"
  76178-966
   Safer Human and Environmental Health
Computer/Technology Tables, All Welded, FlipTop, AmTab
  76178-966
   Safer Human and Environmental Health
Computer/Technology Tables, All Welded, FlipTop, AmTab
Computer/Technology Tables, All Welded, FlipTop, AmTab

Flip top computer and technology table, 4 - 5, 29", 84×30×29"

 
$5,110.05
   
Each
 
 

All-welded computer tables are used every day throughout the world in multi-purpose media and tech environments. Our high-quality state-of-the-art tables are safe, durable, easy to operate, and a great long-term value. Tables are perfect for heavy-use and heavy-turn settings providing years of trusted service.

  • Affordable long-term value
  • 100% made in the USA
  • 15 year warranty

Custom high-speed gas cylinder stabilizer assist for long-lasting opening and closing functions and much more.

 : LEED 2009 certified, ANSI/BIFMA x7.1 certified, UL certified.

 
  • No. of Users:
    4 - 5
  • Désignation:
    Flip top computer and technology table
  • Hauteur:
    29"
  • W×D×H:
    84×30×29"
  • Cat. No.:
    76178-966
  • Environmental certification:
    MAS GREEN
  • Environmentally Preferable:
    Safer Human and Environmental Health