TRIS BUFFER 0.1M 30ML CS/12
Catalog # MSPP-BT103
Supplier: CHARLES RIVER ENDOSAFE MS
TRIS BUFFER 0.1M 30ML CS/12
Catalog # MSPP-BT103
Supplier: CHARLES RIVER ENDOSAFE MS
Supplier Number: BT103
Frequently Bought Together
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Supplier: Air-Tite
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Preserved Starfish
Supplier: Avantor
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Tetrahydrofuran ≥99.9% (by GC) unstabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Acide sulfurique 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Méthanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Sodium hydroxide 98%, pellets
Supplier: Thermo Scientific Chemicals
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
EDTA 0.800 M, cartridge for digital titrator
Supplier: Hach
Accessories for Gas Regulators
Supplier: VWR International
Iodine 0.01 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International