Motic BA210E LED Compound Microscopes
BA210E Phase package
Catalog # 76163-614
Supplier:   VWR International
Motic BA210E LED Compound Microscopes
Catalog # 76163-614
Supplier:  VWR International
Motic BA210E LED Compound Microscopes
Motic BA210E LED Compound Microscopes
BA210E Phase package
Price
1 412,15 $
Vente
Valable jusqu'au 31/12/9999
Log in pour la disponibilité
Each
Product Details & Documents
The BA210Elite model introduces superior improvements both in optical and mechanical performance. Meant to be used in Educational life sciences, Medical and a variety of Biological applications, the BA210E is designed for ease of use and longevity due to Motic's complete understanding of the educational market.
  • Widefield high eyepoint eyepiece N-WF10×/20 mm with diopter
  • Adjustment on both eyepieces, with rubber eyecup (paired)
  • Reversed quadruple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment
  • Built in low position coaxial mechanical stage (right-hand control)
  • Focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot
The BA210E model introduces student-proof features both in optical and mechanical aspects. The new rackless stage concept without prominent gear rack allows even more convenient use of the x/y movement, while increasing user safety in teaching situations.
For full freedom of illumination options, the new model BA210E has an easy interchangeability of its 6 V/30 W Halogen bulb with LED modules of different colour temperatures (4500K, 6000K). Extended life-time of LEDs here is combined with cost saving aspects. Light consuming contrast methods like Phase contrast, Polarization or Dark field can easily performed.
 : Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
 : The BA210E Phase package option (76163-614) includes upright compound microscope, trinocular head, widefield high eyepoint 10× eyepieces, CCIS EC Plan achromat/phase objectives (PL4×, PL100× S-Oil, Ph10X), rackless stage, NA 1.25 abbe condenser, phase slider, phase centering telescope, 3WLED module.
Spécifications
  • Désignation:
    BA210E Phase package