Ward’s® Adjustable Volume Micropipettes
20 μL Micropipette, 2 - 20 µl, 0.5 µl
Catalog # 470231-608
Supplier: DLAB SCIENTIFIC INC.
: 7010301005
Jump to:
Product Details & Documents
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
- Fully autoclavable
- Slim, lightweight ergonomic design
- Compatible with most universal tip brands
- Tool included for quick and easy recalibration
- Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Spécifications
- Volume:2 - 20 µl
- Désignation:20 μL Micropipette
- Increments:0.5 µl