Mix & Go!™ E. coli Transformation Kit and Buffer Set | Avantor
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Mix & Go!™ E. coli transformation kit, up to 20 ml
Catalog # 77002-226
Supplier:   Zymo Research
Fournisseur #:  T3001
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Catalog # 77002-226
Supplier:  Zymo Research
CAS Number:  
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Mix & Go!™ E. coli transformation kit, up to 20 ml
CAS Number:  
Price
$220.68
Log in pour la disponibilité
Each Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The Mix & Go!™ E. coli transformation kit and Mix & Go!™ E. coli transformation buffer set are convenient methods for the preparation of competent E. coli cells for simple and highly efficient DNA transformation.
  • It is very simple and efficient DNA transformation
  • It completely eliminates the requirement for heat shocking and related procedures
  • Transformation efficiencies are typically on the order of 10to 109 transformants/µg plasmid DNA
The Mix & Go! method completely eliminates the requirement for heat shocking and related procedures. Instead, Mix & Go! bacterial transformation can be performed by adding DNA to Mix & Go! competent cells and spreading onto a plate. Transformation efficiencies are typically on the order of 10to 109 transformants/µg plasmid DNA with most E. coli strains. These formulated reagents make it easy to generate Mix & Go! competent cells from current E. coli strains that are available in the laboratory. Simply grow the E. coli strain of your choice, wash, then resuspend the cells in the provided buffers. The cells are now transformation ready. The Mix & Go! E. coli Transformation Kit includes all buffers and ZymoBroth medium to generate 20 ml of Mix & Go! competent cells. The Mix & Go! E. coli Transformation Buffer Set includes all buffers that are required to generate 60 ml of Mix & Go! competent cells, and the medium (broth) is supplied by the user.
Spécifications
Spécifications
  • Désignation:
    Mix & Go!™ E. coli transformation kit
  • Taille:
    up to 20 ml