Quick-DNA/RNA™ Miniprep Plus Kits
Quick-DNA/RNA™ Miniprep Plus, for Cells, any tissues, blood, biological fluids, 50 Preps
Catalog # 77001-854
Supplier: Zymo Research
Fournisseur #: D7003
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
Spécifications
Spécifications
- Désignation:Quick-DNA/RNA™ Miniprep Plus
- Taille:50 Preps
- Details:for Cells, any tissues, blood, biological fluids
- Cat. No.:77001-854
- Binding capacity:100 µg
- Yield:100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)