VWR® Cleanroom Clogs | Avantor
VWR® Cleanroom Clogs
XS (US: 4–5.5; EU: 35–36), White
Catalog # 76077-098
Supplier:   VWR International
VWR® Cleanroom Clogs
Catalog # 76077-098
Supplier:  VWR International
VWR® Cleanroom Clogs
VWR® Cleanroom Clogs
XS (US: 4–5.5; EU: 35–36), White
Price
$272.06
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pair Case of 6
Product Details & Documents
Cleanroom Clogs from VWR® provide comfort and durability for use in cleanroom environments.
  • Antistatic
  • Autoclavable
  • Smooth surface for day long comfort
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Non-slip sole
  • Anatomic heel basin
Cleanroom Clogs from VWR® have been designed with foot comfort in mind. Smooth surface and aeration holes on both sides provide day long comfort. Special conical design prevents spilled liquids from entering inside the clog.
These shoes are suitable for autoclave sterilization. They can also be cleaned in 100 °C with domestic detergents, liquid disinfecting solutions, EO, or formaldehyde.
Ideal for use with VWR® Cleanroom Disposable or Launderable Socks.
Ordering information: Clogs can be ordered by the pair or per case.
Packaging: Packaged six pair per case.
Spécifications
  • Pointure:
    XS (US: 4–5.5; EU: 35–36)
  • Color:
    White