Thread Adapters, Polypropylene
External thread: 2" coarse Tri-Sure - 64 mm (2.52") (BSI), Brown
Catalog # 76077-158
Supplier: BURKLE INC
Supplier #: 0600-3002
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Specifications
- Color:Brown
- Désignation:External thread: 2" coarse Tri-Sure - 64 mm (2.52") (BSI)