Thread Adapters, Polypropylene
External thread: 61 mm (2.40") - 2" BSP, Black
76077-154
Supplier #: 0600-3007
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
- Color:Black
- Désignation:External thread: 61 mm (2.40") - 2" BSP