Thread Adapters, Polypropylene
Internal thread: 63 mm (2.48") - 2" BSP, White
Catalog # 76077-144
Supplier: BURKLE INC
Supplier #: 0600-2006
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Specifications
- Color:White
- Désignation:Internal thread: 63 mm (2.48") - 2" BSP