Thread Adapters, Polypropylene
External thread: 64 mm (2.52") (BSI), Internal thread: DIN 71, Brown/blue
  76077-136
 :   BURKLE INC
 :  0600-1001
Thread Adapters, Polypropylene
  76077-136
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene
External thread: 64 mm (2.52") (BSI), Internal thread: DIN 71, Brown/blue
$97.05
Each
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
  • Color:
    Brown/blue
  • Désignation:
    External thread: 64 mm (2.52") (BSI), Internal thread: DIN 71