EverMark Select Microscope Slides, Azer Scientific | Avantor
EverMark Select Microscope Slides, Azer Scientific
Frosted adhesive slides, color-coded, Aqua, 1.0 mm, 75×25 mm (3×1")
Catalog # 89033-063
EverMark Select Microscope Slides, Azer Scientific
Catalog # 89033-063
Supplier:  
EverMark Select Microscope Slides, Azer Scientific
EverMark Select Microscope Slides, Azer Scientific
Frosted adhesive slides, color-coded, Aqua, 1.0 mm, 75×25 mm (3×1")
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR SLIDE VISTA AQUA PK72 Catalog #16005-108
Le produit 16005-108 est similaire à 89033-063 mais peut avoir un fabricant et/ou un emballage différents.
Product Details & Documents
Made from high-quality glass, these slides are pre-cleaned with smooth grounded edges.
  • High-quality glass
  • Pre-cleaned
  • Smooth, grounded edges
  • Single-side, color-frosted ends
  • Clipped corners
  • Chemical-resistant
All slides have single-side, color-frosted ends to help avoid sticking and jamming
Adhesive slides feature a permanent positive charge to firmly anchor all types of specimens to the slide. Slides with clipped corners are approved for use with Leica™, Sakura™, Sysmex™, and other automated instruments.
Frosted finish is resistant to common lab chemicals.
Spécifications
  • Color:
    Aqua
  • Désignation:
    Frosted adhesive slides, color-coded
  • L×l:
    75×25 mm (3×1")
  • Épaisseur:
    1.0 mm