$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
VWR DRAWERRACK4X4FOR 100-P SLIDE BOXES
76053-196
:
CRYSTAL TECHNOLOGY & INDUSTRIES
VWR DRAWERRACK4X4FOR 100-P SLIDE BOXES
76053-196
:
CRYSTAL TECHNOLOGY & INDUSTRIES
:
UFDS-100-44
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH