Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VWR UPRIGHT RACK2X5X2 FOR 50C 0.5ML BOX
76052-466
:
CRYSTAL TECHNOLOGY & INDUSTRIES
VWR UPRIGHT RACK2X5X2 FOR 50C 0.5ML BOX
76052-466
:
CRYSTAL TECHNOLOGY & INDUSTRIES
:
UFM-2505
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH