$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
VWR UPRIGHT RACK7X24 FOR 96/384-W PLATES
76051-744
:
CRYSTAL TECHNOLOGY & INDUSTRIES
VWR UPRIGHT RACK7X24 FOR 96/384-W PLATES
76051-744
:
CRYSTAL TECHNOLOGY & INDUSTRIES
:
UFMP-716L
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH