VWR® Vertical Racks
VWR® Vertical Racks
Vertical freezer rack, 6× 3" boxes, 5½", 19⅜", 5⅝"
Catalog # 76051-660
Supplier:   VWR International
VWR® Vertical Racks
Catalog # 76051-660
Supplier:  VWR International
VWR® Vertical Racks
VWR® Vertical Racks
Vertical freezer rack, 6× 3" boxes, 5½", 19⅜", 5⅝"
Price
$111.31
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Vertical racks for 3" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Specifications
  • Profondeur:
    5½"
  • Largeur:
    5⅝"
  • Capacité:
    6× 3" boxes
  • Désignation:
    Vertical freezer rack
  • Hauteur:
    19⅜"