VWR® Vertical Racks | Avantor
VWR® Vertical Racks
Vertical rack, 12× 2" mini boxes, 3¼", 26⅝", 3⅜"
Catalog # 76051-574
Supplier:   VWR International
VWR® Vertical Racks
Catalog # 76051-574
Supplier:  VWR International
VWR® Vertical Racks
VWR® Vertical Racks
Vertical rack, 12× 2" mini boxes, 3¼", 26⅝", 3⅜"
Price
$104.71
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Vertical freezer racks for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Spécifications
  • Profondeur:
    3¼"
  • Largeur:
    3⅜"
  • Capacité:
    12× 2" mini boxes
  • Désignation:
    Vertical rack
  • Hauteur:
    26⅝"