Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Specific Gravity Cubes

Specific Gravity Cubes

Cube iron, 20 mm
Catalog # 470230-028
Supplier:   Wards
Supplier #:  PH0108F
Specific Gravity Cubes
Catalog # 470230-028
Supplier:  Wards
Specific Gravity Cubes
Specific Gravity Cubes

Cube iron, 20 mm

Price
$10.90
Log in for availability
Each
Product Details & Documents

Specific Gravity Cubes with Hooks.

  • Brass, iron, aluminum and zinc 20mm cubes
  • Hooks

Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.

Specifications
  • Désignation:
    Cube iron, 20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.