Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Specific Gravity Cubes | Avantor

Specific Gravity Cubes

Cube al, 20 mm
Catalog # 470230-026
Supplier:   Wards
Fournisseur #:  PH0108E
Specific Gravity Cubes
Catalog # 470230-026
Supplier:  Wards
Specific Gravity Cubes
Specific Gravity Cubes

Cube al, 20 mm

Price
$11.40
Log in pour la disponibilité
Each
Product Details & Documents

Specific Gravity Cubes with Hooks.

  • Brass, iron, aluminum and zinc 20mm cubes
  • Hooks

Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.

Spécifications
  • Désignation:
    Cube al, 20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.