Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Specific Gravity Cubes

Cube brass, 20 mm
Catalog # 470230-022
Supplier:   Wards
 :  PH0108A
Specific Gravity Cubes
Catalog # 470230-022
Supplier:  Wards
Specific Gravity Cubes
Specific Gravity Cubes

Cube brass, 20 mm

Price
$13.45
Log in pour la disponibilité
Each
Product Details & Documents

Specific Gravity Cubes with Hooks.

  • Brass, iron, aluminum and zinc 20mm cubes
  • Hooks

Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.

Spécifications
  • Désignation:
    Cube brass, 20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.