Specific Gravity Cubes | Avantor
Specific Gravity Cubes
Specific Gravity Cubes
Catalog # 470230-020
Supplier:   Wards
Fournisseur #:  PH0108
Specific Gravity Cubes
Catalog # 470230-020
Supplier:  Wards
Specific Gravity Cubes
Specific Gravity Cubes
Specific Gravity Cubes
Liquidation
$21.35
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
Specific Gravity Cubes with Hooks.
  • Brass, iron, aluminum and zinc 20mm cubes
  • Hooks
Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.
Spécifications
  • Désignation:
    Specific Gravity Cubes