Specific Gravity Cubes
Specific Gravity Cubes
Catalog # 470230-020
Supplier: Wards
Fournisseur #: PH0108
Jump to:
Product Details & Documents
Specific Gravity Cubes with Hooks.
- Brass, iron, aluminum and zinc 20mm cubes
- Hooks
Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.
Spécifications
- Désignation:Specific Gravity Cubes