U-2D™ Micro-sample management system with racked inserts | Avantor
U-2D™ Micro-sample management system with racked inserts
U-2D™ replacement inserts, low volume, glass. Clear, 500 µl with a wide, conical point
Catalog # 76057-654
Fournisseur #:  95296-I-500
U-2D™ Micro-sample management system with racked inserts
Catalog # 76057-654
U-2D™ Micro-sample management system with racked inserts
U-2D™ Micro-sample management system with racked inserts
U-2D™ replacement inserts, low volume, glass. Clear, 500 µl with a wide, conical point
Price
$197.00
Log in pour la disponibilité
Pack of 100
Product Details & Documents
Manage multiple low volume samples in a 96 well format and a unique split rack with glass inserts for vials.
  • 350 µl inserts can be used individually by dropping them into a 2 ml vial
  • Compatible with plate readers
  • Fill the inserts with multi-channel pipettors for quick and easy sample preparation
  • Transport inserts safely
Use this unique sample management system that is designed for preparing samples or for injecting them with autosamplers. Each rack is supplied with 96 glass inserts that are installed and ready for use. The included insert rack can be split from the base allowing full visual inspection of the insert bottoms to identify and remove bubble formation. Achieve even and thorough thermal control of all samples by inserting into a water bath or using other heat or cooling methods. The complete rack and inserts are compatible with all autosamplers that accept the ANSI standard 96-well format if desired.
Spécifications
  • Désignation:
    U-2D™ replacement inserts, low volume, glass. Clear, 500 µl with a wide, conical point
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Zone Free Sealing Film for U-2D™ Sealing Film
    Zone Free Sealing Film for U-2D™ Sealing Film
    10002-828
    Log in pour la disponibilité
    Case of 50 - $75.97
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent Black Marker, for use with U-2D™ Plate System
    Permanent Black Marker, for use with U-2D™ Plate System
    10002-812
    Log in pour la disponibilité
    Each - $8.01
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent Red Marker, for use with U-2D™ Plate System
    Permanent Red Marker, for use with U-2D™ Plate System
    10002-816
    Log in pour la disponibilité
    Each - $7.62
Informations sur le produit
  • Catalog #
    Availability
    Price
  • U-2D™ plate system easy purchase pack with rack and base only. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical points
    U-2D™ plate system easy purchase pack with rack and base only. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical points
    10014-946
    Log in pour la disponibilité
    Each - $201.25
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent Red Marker, for use with U-2D™ Plate System
    Permanent Red Marker, for use with U-2D™ Plate System
    10002-818
    Log in pour la disponibilité
    Pack of 10 - $57.86
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent Silver Marker, for use with U-2D™ Plate System, Extra Fine Tip
    Permanent Silver Marker, for use with U-2D™ Plate System, Extra Fine Tip
    75812-810
    Log in pour la disponibilité
    Each - $20.54
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent Black Marker, for use with U-2D™ Plate System, 0.45mm Fine Tip
    Permanent Black Marker, for use with U-2D™ Plate System, 0.45mm Fine Tip
    75812-800
    Log in pour la disponibilité
    Each - $19.67
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Paddles for U-2D™ Sealing Film
    Paddles for U-2D™ Sealing Film
    10002-820
    Log in pour la disponibilité
    Pack of 5 - $32.67
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent Black Marker, for use with U-2D™ Plate System
    Permanent Black Marker, for use with U-2D™ Plate System
    10002-814
    Log in pour la disponibilité
    Pack of 10 - $57.86
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Roller for U-2D™ Sealing Film
    Roller for U-2D™ Sealing Film
    10002-822
    Log in pour la disponibilité
    Each - $113.36
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Permanent White Marker, for use with U-2D™ Plate System, Extra Fine Tip
    Permanent White Marker, for use with U-2D™ Plate System, Extra Fine Tip
    75812-812
    Log in pour la disponibilité
    Each - $20.54