Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Biofluid & Cell Buffer (Red), Zymo Research

Biofluid & Cell Buffer (Red), Zymo Research

45 mL
Catalog # 77001-352
Supplier:   Zymo Research
Supplier #:  D4068-1-45
Biofluid & Cell Buffer (Red), Zymo Research
Catalog # 77001-352
Supplier:  Zymo Research
CAS Number:  
Biofluid & Cell Buffer (Red), Zymo Research
Biofluid & Cell Buffer (Red), Zymo Research

45 mL

CAS Number:  
Price
$146.34
Log in for availability
Each (45ml)
Minimum order quantity is 1
Product Details & Documents
Specifications

Specifications

  • Taille:
    45 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.