pH Test Papers
pH Test Papers, Litmus Blue ACS (12 Vials)
Catalog # 470123-117
Supplier: Precision Laboratories
Supplier #: 180-12V-100
Jump to:
Product Details & Documents
Each stoppered vial contains 100 (1.8 x 2") strips.
Each stoppered vial contains 100 (1.8" x 2") strips.
Specifications
- Désignation:pH Test Papers, Litmus Blue ACS (12 Vials)