Extracta™ DNA Prep for PCR, Quantabio
Product CA76047-142 is a replacement for 76047-142 and has the same packaging and specifications, but may be a different quantity.
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Extracta DNA Prep for PCR is a two-component reagent kit for rapid extraction of PCR-ready genomic DNA from a variety of tissues. Samples are processed in less than 30 minutes with minimal hands-on time and technical skill.
- Simple, fast protocol
- Compatible with laboratory automation
- DNA is suitable for qPCR, High Resolution Melt Analysis, endpoint PCR and other applications
- Works with wide range of tissue including hair, buccal swabs and mouse tails
- Optional 10 minute protocol for high-throughput applications
Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.
The Extracta™ DNA prep for PCR consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva. The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.
Ordering information: DNA Prep kit includes extraction reagent and stabilization buffer.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Désignation:Extracta™ DNA prep for PCR
- Taille:2.5 ml
- Cat. No.:76047-142
- Prep size:Scalable
- Refine target molecule:Total DNA
- Target molecule:DNA
- Method / Format:Extraction for direct PCR
- Description:Extracta™ DNA prep for PCR
- Sample type:Cultured cells|Human/animal cells and tissues
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



