Neodymium Disk Magnet
Neodymium Disk Magnet, 0.5" Diameter, 0.125 " Thick
Catalog # 470229-350
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
NEODYMIUM DISK 0.5 INCH BY .25 INCH PK3 Catalog #470229-348
Le produit 470229-348 est similaire à 470229-350 mais le fabricant et les spécifications peuvent varier.
Product Details & Documents
A powerful magnet for a variety of experiments.
- Very high resistance to demagnetization
- High energy for size
- Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. All of the magnets have a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).
Spécifications
- Désignation:Neodymium Disk Magnet, 0.5" Diameter, 0.125 " Thick