Neodymium Disk Magnet
Neodymium Disk Magnet, 0.5" Diameter, 0.25 " Thick
Catalog # 470229-348
Supplier: Master Magnetics
: ND143N-35BXX3
Jump to:
Product Details & Documents
A powerful magnet for a variety of experiments.
- Very high resistance to demagnetization
- High energy for size
- Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. All of the magnets have a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).
Spécifications
- Désignation:Neodymium Disk Magnet, 0.5" Diameter, 0.25 " Thick



