Crystallizing Dish
100 ml, 70 mm, 40 mm
Catalog # 470229-210
Supplier: Wards
Fournisseur #: CH0066E
Jump to:
Product Details & Documents
Ideal for storage and crystallization.
- Borosilicate glass
- Flat bottom
- Easy pour spout
Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.
Spécifications
- Ø:70 mm
- Capacité:100 ml
- Hauteur:40 mm