Engineering Hydraulic Syringes
60 mL Superlock Syringe
Catalog # 470229-344
Supplier: Avantor
Jump to:
Product Details & Documents
Perfect for Makerspace and hydraulic projects.
Convenient packs of 10 for use in a variety of Makerspace and engineering challenges. All come with a lure lock fitting that fits snuggly into vinyl tubing, but can not be fitted with a needle.
Spécifications
- Désignation:60 mL Superlock Syringe