Engineering Hydraulic Syringes | Avantor
Engineering Hydraulic Syringes
60 mL Superlock Syringe
Catalog # 470229-344
Supplier:   Avantor
Engineering Hydraulic Syringes
Catalog # 470229-344
Supplier:  Avantor
Engineering Hydraulic Syringes
Engineering Hydraulic Syringes
60 mL Superlock Syringe
Price
$55.99
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Perfect for Makerspace and hydraulic projects.
Convenient packs of 10 for use in a variety of Makerspace and engineering challenges. All come with a lure lock fitting that fits snuggly into vinyl tubing, but can not be fitted with a needle.
Spécifications
  • Désignation:
    60 mL Superlock Syringe