Engineering Hydraulic Syringes
30 mL Superlock Syringe
470229-342
: Avantor
Perfect for Makerspace and hydraulic projects.
Convenient packs of 10 for use in a variety of Makerspace and engineering challenges. All come with a lure lock fitting that fits snuggly into vinyl tubing, but can not be fitted with a needle.
- Désignation:30 mL Superlock Syringe