Engineering Hydraulic Syringes
30 mL Superlock Syringe
  470229-342
 :   Avantor
Engineering Hydraulic Syringes
  470229-342
 :  Avantor
Engineering Hydraulic Syringes
Engineering Hydraulic Syringes
30 mL Superlock Syringe
$37.79
Pack of 10
Perfect for Makerspace and hydraulic projects.
Convenient packs of 10 for use in a variety of Makerspace and engineering challenges. All come with a lure lock fitting that fits snuggly into vinyl tubing, but can not be fitted with a needle.
  • Désignation:
    30 mL Superlock Syringe