Fossil Study Packs
Le produit 470145-998 est un remplacement de 470228-884 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Product Details & Documents
These packs feature representative fossils from the phanerozoic eon.
Representative fossils from the phanerozoic eon
- Marine and terrestrial vertebrates, invertebrates, and plants
-
Nine major fossil groups are represented
Bundle and save with the complete set!
Each study pack contains 10 of each selected fossil along with key information about its characteristics and important place in earth history.
9 major fossil groups are represented, spanning from the cambrian to tertiary periods. Packs include marine vertebrates, invertebrates, and plants.
Spécifications
- Désignation:Trilobites