ProPette™ Electronic Pipette Controller
Lithium Battery
Catalog # 470228-482
Supplier: MTC BIO, INC.
Supplier #: P6080-BA
Jump to:
Product Details & Documents
ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.
- QuickStand™ prevents pipette contamination
- Variable speed control
- Cordless and rechargable
- Provides 8 hours of continuous operation
- Fits serological pipettes from 1 to 100 ml
- Ambidexterous
Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.
Specifications
- Désignation:Lithium Battery