ProPette™ Electronic Pipette Controller
Pipette Controller, with Euro 2-Prong Plug
Catalog # 470228-480
Supplier: MTC BIO, INC.
Fournisseur #: P6080-I
Jump to:
Product Details & Documents
ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.
- QuickStand™ prevents pipette contamination
- Variable speed control
- Cordless and rechargable
- Provides 8 hours of continuous operation
- Fits serological pipettes from 1 to 100 ml
- Ambidexterous
Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.
Spécifications
- Désignation:Pipette Controller, with Euro 2-Prong Plug