ProPette™ Electronic Pipette Controller | Avantor
ProPette™ Electronic Pipette Controller
Pipette Controller, with US Plug
Catalog # 470228-478
Supplier:   MTC BIO, INC.
Fournisseur #:  P6080
ProPette™ Electronic Pipette Controller
Catalog # 470228-478
Supplier:  MTC BIO, INC.
ProPette™ Electronic Pipette Controller
ProPette™ Electronic Pipette Controller
Pipette Controller, with US Plug
Price
$607.55
Log in pour la disponibilité
Each
Product Details & Documents
ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.
  • QuickStand™ prevents pipette contamination
  • Variable speed control
  • Cordless and rechargable
  • Provides 8 hours of continuous operation
  • Fits serological pipettes from 1 to 100 ml
  • Ambidexterous
Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.
Spécifications
  • Désignation:
    Pipette Controller, with US Plug